Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 55142327-55142497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFR
Entrez Gene ID
1956
Gene Name
epidermal growth factor receptor
Gene Aliases
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA
UniGene
Hs.488293
Gene Chromosome Location
Chr.7: 55019032-55207338 on Build GRCh38
Transcripts
NM_005228.3 NM_201282.1 NM_201283.1 NM_201284.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6970485 c.188G>A p.G63E Chr.7: 55142385-55142385 on Build GRCh38
6652263 c.180G>T p.V60V Chr.7: 55142377-55142377 on Build GRCh38
5730403 c.185T>G p.L62R Chr.7: 55142382-55142382 on Build GRCh38
3266511 c.203delC p.Y69fs*11 Chr.7: 55142400-55142400 on Build GRCh38
3639717 c.139C>T p.H47Y Chr.7: 55142336-55142336 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778638117 Chr.7: 55142423-55142423 on Build GRCh38
rs760228905 Chr.7: 55142372-55142372 on Build GRCh38
rs763572530 Chr.7: 55142373-55142373 on Build GRCh38
rs762825566 Chr.7: 55142467-55142467 on Build GRCh38
rs767259994 Chr.7: 55142364-55142364 on Build GRCh38
rs780215578 Chr.7: 55142466-55142466 on Build GRCh38
rs369580836 Chr.7: 55142409-55142409 on Build GRCh38
rs752438083 Chr.7: 55142371-55142371 on Build GRCh38
rs759219499 Chr.7: 55142355-55142355 on Build GRCh38
rs758238090 Chr.7: 55142463-55142463 on Build GRCh38
rs750239730 Chr.7: 55142446-55142446 on Build GRCh38
rs200383389 Chr.7: 55142419-55142419 on Build GRCh38
rs61731794 Chr.7: 55142428-55142428 on Build GRCh38

View Full Product Details