Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41747385-41747557 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6918450 c.295C>T p.R99W Chr.4: 41747483-41747483 on Build GRCh38
6981393 c.298C>T p.R100C Chr.4: 41747480-41747480 on Build GRCh38
6980376 c.272G>T p.G91V Chr.4: 41747506-41747506 on Build GRCh38
174667 c.271G>A p.G91S Chr.4: 41747507-41747507 on Build GRCh38
6745584 c.299G>A p.R100H Chr.4: 41747479-41747479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766200980 Chr.4: 41747525-41747525 on Build GRCh38
rs988876214 Chr.4: 41747467-41747467 on Build GRCh38
rs956551152 Chr.4: 41747455-41747455 on Build GRCh38
rs747055965 Chr.4: 41747433-41747433 on Build GRCh38
rs199611260 Chr.4: 41747469-41747469 on Build GRCh38
rs753545181 Chr.4: 41747529-41747529 on Build GRCh38
rs763804374 Chr.4: 41747463-41747463 on Build GRCh38
rs762806075 Chr.4: 41747459-41747459 on Build GRCh38
rs560843362 Chr.4: 41747405-41747405 on Build GRCh38
rs749994532 Chr.4: 41747511-41747511 on Build GRCh38
rs755664325 Chr.4: 41747513-41747513 on Build GRCh38
rs776824229 Chr.4: 41747456-41747456 on Build GRCh38
rs996060917 Chr.4: 41747517-41747517 on Build GRCh38
rs104893855 Chr.4: 41747479-41747479 on Build GRCh38
rs201892150 Chr.4: 41747490-41747490 on Build GRCh38
rs112718633 Chr.4: 41747436-41747436 on Build GRCh38
rs528408156 Chr.4: 41747510-41747510 on Build GRCh38
rs140002346 Chr.4: 41747472-41747472 on Build GRCh38

View Full Product Details