Assay Design



Gene Transcripts

Gene Symbol
GTPBP1
Entrez Gene ID
9567
Gene Name
GTP binding protein 1
Gene Aliases
GP-1, GP1, HSPC018
Chromosome Location
Chr.22: 38704561 - 38738265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004286.4 NP_004277.2 12 - 12 4819 104
GenBank mRNA AF077204.1 - 1 - 1 449 103
AF370385.1 - 1 - 1 2392 104
AK095950.1 - 1 - 1 2380 104
BC001701.2 - 1 - 1 336 104
BC046350.2 - 1 - 1 330 104

View Full Product Details