Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199140.1 | NP_001186069.1 | 8 - 8 | 3716 | 103 |
NM_031445.2 | NP_113633.2 | 8 - 8 | 3688 | 103 | |
XM_005263804.4 | XP_005263861.1 | 9 - 9 | 4319 | 103 | |
XM_005263806.4 | XP_005263863.1 | 8 - 8 | 4208 | 103 | |
XM_005263807.4 | XP_005263864.1 | 7 - 7 | 4073 | 103 | |
XM_011511952.2 | XP_011510254.1 | 8 - 8 | 4209 | 103 | |
XM_011511954.2 | XP_011510256.1 | 9 - 9 | 4296 | 103 | |
XM_011511955.2 | XP_011510257.1 | 7 - 7 | 3238 | 103 | |
GenBank mRNA | AK024993.1 | - | 1 - 1 | 1163 | 103 |
AK026685.1 | - | 1 - 1 | 2049 | 103 | |
BC016181.1 | - | 8 - 8 | 3688 | 103 | |
BC078174.1 | - | 8 - 8 | 3698 | 103 | |
BC084552.1 | - | 1 - 1 | 36 | 103 |