Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032444.2 | NP_115820.2 | 14 - 15 | 5782 | 61 |
XM_011522715.2 | XP_011521017.1 | 14 - 15 | 5793 | 61 | |
XM_017023775.1 | XP_016879264.1 | 11 - 12 | 5187 | 61 | |
GenBank mRNA | AB058687.1 | - | - | 3156 | 61 |
AK095411.1 | - | 3 - 4 | 1652 | 61 | |
AL442083.1 | - | 1 - 2 | 322 | 61 | |
BC036335.1 | - | 5 - 6 | 3174 | 61 |