Assay Design



Gene Transcripts

Gene Symbol
MYOM3
Entrez Gene ID
127294
Gene Name
myomesin 3
Gene Aliases
-
Chromosome Location
Chr.1: 24056041 - 24112175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152372.3 NP_689585.3 33 - 34 4100 60
GenBank mRNA AF086254.1 - 2 - 3 190 60
AK093280.1 - 6 - 7 986 60
AK128233.1 - 25 - 26 3160 60
BC014671.1 - 7 - 8 620 60
BX537911.1 - 32 - 33 3824 60
BX647453.1 - 33 - 34 4061 60
BX647455.1 - 33 - 34 4100 60
BX647644.1 - 33 - 34 4061 60

View Full Product Details