Assay Design
Gene Transcripts
- Gene Symbol
- AMZ2P1
- Entrez Gene ID
-
201283
- Gene Name
- archaelysin family metallopeptidase 2 pseudogene 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 64966550 - 64975585 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026903.1 |
|
1 - 2 |
1031 |
100 |
GenBank mRNA |
AK056627.1 |
- |
1 - 2 |
1014 |
100 |
BX537538.1 |
- |
1 - 2 |
905 |
100 |
View Full Product Details