Assay Design



Gene Transcripts

Gene Symbol
FBXO39
Entrez Gene ID
162517
Gene Name
F-box protein 39
Gene Aliases
CT144, Fbx39
Chromosome Location
Chr.17: 6776184 - 6798929 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153230.2 NP_694962.1 2 - 3 1143 77
XM_011523697.1 XP_011521999.1 2 - 3 1219 77
GenBank mRNA BC034782.1 - 2 - 3 1123 77

View Full Product Details