Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260497.1 | NP_001247426.1 | 1 - 2 | 536 | 69 |
NM_001260498.1 | NP_001247427.1 | 1 - 2 | 536 | 69 | |
NM_001260499.1 | NP_001247428.1 | 1 - 2 | 536 | 69 | |
NM_139136.3 | NP_631874.1 | 1 - 2 | 536 | 69 | |
NM_139137.3 | NP_631875.1 | 1 - 2 | 536 | 69 | |
NM_153748.2 | NP_715624.1 | 1 - 2 | 536 | 69 | |
XM_005268855.3 | XP_005268912.1 | 1 - 2 | 592 | 69 | |
XM_006719382.3 | XP_006719445.1 | 1 - 2 | 592 | 69 | |
XM_006719383.3 | XP_006719446.1 | 1 - 2 | 592 | 69 | |
XM_006719384.3 | XP_006719447.1 | 1 - 2 | 603 | 69 | |
XM_006719385.3 | XP_006719448.1 | 1 - 2 | 592 | 69 | |
XM_006719390.3 | XP_006719453.1 | 1 - 2 | 592 | 69 | |
XM_017019282.1 | XP_016874771.1 | 1 - 2 | 592 | 69 | |
GenBank mRNA | AB209994.1 | - | 1 - 2 | 235 | 69 |
AK094720.1 | - | 1 - 2 | 519 | 69 | |
AK294269.1 | - | 1 - 2 | 248 | 69 | |
AK309245.1 | - | 1 - 2 | 223 | 69 | |
BC093635.1 | - | 1 - 2 | 70 | 69 | |
BC111991.1 | - | 1 - 2 | 70 | 69 | |
DC323864.1 | - | 1 - 2 | 536 | 69 |