Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289050.1 | NP_001275979.1 | 8 - 9 | 1384 | 70 |
NM_016609.4 | NP_057693.3 | 9 - 10 | 1776 | 70 | |
NM_020372.3 | NP_065105.2 | 8 - 9 | 1563 | 70 | |
NR_110290.1 | 7 - 8 | 1441 | 70 | ||
XM_005267747.4 | XP_005267804.1 | 9 - 10 | 3906 | 70 | |
XM_005267748.4 | XP_005267805.1 | 7 - 8 | 3531 | 70 | |
XM_017021361.1 | XP_016876850.1 | 9 - 10 | 3903 | 70 | |
XM_017021362.1 | XP_016876851.1 | 9 - 10 | 3847 | 70 | |
GenBank mRNA | AJ243653.1 | - | 6 - 7 | 824 | 70 |
BC020565.1 | - | 8 - 9 | 1382 | 70 | |
BC090870.1 | - | 8 - 9 | 1060 | 70 | |
BC111015.1 | - | 7 - 8 | 923 | 70 | |
BX161416.1 | - | 9 - 10 | 1735 | 70 | |
BX647316.1 | - | 5 - 6 | 1131 | 70 | |
DQ658848.1 | - | 8 - 9 | 1154 | 70 |