Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018298.10 | NP_060768.8 | 2 - 3 | 378 | 113 |
XM_005271003.1 | XP_005271060.1 | 3 - 4 | 775 | 113 | |
XM_006710750.1 | XP_006710813.1 | 3 - 4 | 594 | 113 | |
XM_011541739.2 | XP_011540041.2 | 2 - 3 | 302 | 113 | |
XM_011541741.2 | XP_011540043.1 | 2 - 3 | 309 | 113 | |
XM_011541742.1 | XP_011540044.1 | 2 - 3 | 353 | 113 | |
GenBank mRNA | AF475085.1 | - | 2 - 3 | 284 | 113 |
AK292206.1 | - | 2 - 3 | 310 | 113 | |
AK314354.1 | - | 2 - 3 | 338 | 113 | |
BC060765.1 | - | 3 - 4 | 608 | 113 | |
BP358726.1 | - | 2 - 3 | 378 | 113 | |
BX647497.1 | - | 3 - 4 | 372 | 113 |