Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201484.1 | NP_001188413.1 | 9 - 10 | 1889 | 74 |
NM_032634.3 | NP_116023.2 | 7 - 8 | 3042 | 74 | |
NM_152850.3 | NP_690577.2 | 8 - 9 | 1437 | 74 | |
XM_005251619.3 | XP_005251676.1 | 7 - 8 | 2667 | 74 | |
XM_017015222.1 | XP_016870711.1 | 8 - 9 | 3159 | 74 | |
XM_017015223.1 | XP_016870712.1 | 8 - 9 | 1818 | 74 | |
XM_017015224.1 | XP_016870713.1 | 8 - 9 | 2082 | 74 | |
GenBank mRNA | AK074064.1 | - | 3 - 4 | 3647 | 74 |
AL833956.1 | - | 7 - 8 | 3075 | 74 | |
AY358472.1 | - | 6 - 7 | 2648 | 74 | |
BC001030.2 | - | 5 - 6 | 767 | 74 | |
BC013987.2 | - | 6 - 7 | 853 | 74 | |
BC029271.1 | - | 6 - 7 | 2313 | 74 | |
BC036916.1 | - | 7 - 8 | 1402 | 74 | |
BC065282.1 | - | 9 - 10 | 1560 | 74 |