Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001269043.1 | NP_001255972.1 | 1 - 2 | 103 | 64 |
NM_001269046.1 | NP_001255975.1 | 1 - 2 | 628 | 64 | |
NM_080618.3 | NP_542185.2 | 2 - 3 | 282 | 64 | |
GenBank mRNA | AF336042.1 | - | 1 - 2 | 81 | 64 |
AK128070.1 | - | 1 - 2 | 185 | 64 | |
AK313990.1 | - | 1 - 2 | 116 | 64 | |
BC130486.1 | - | 1 - 2 | 59 | 64 | |
DQ294738.1 | - | 1 - 2 | 553 | 64 | |
DQ778109.1 | - | 2 - 3 | 282 | 64 | |
DQ778111.1 | - | 1 - 2 | 81 | 64 | |
DQ778112.1 | - | 1 - 2 | 628 | 64 |