Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170543.1 | NP_001164014.1 | 3 - 4 | 567 | 75 |
NM_001170544.1 | NP_001164015.1 | 3 - 4 | 567 | 75 | |
NM_138575.3 | NP_612642.2 | 3 - 4 | 567 | 75 | |
GenBank mRNA | AK097688.1 | - | 3 - 4 | 914 | 75 |
BC008196.1 | - | 3 - 4 | 524 | 75 | |
BC047906.2 | - | 2 - 3 | 222 | 75 | |
DR155760.1 | - | 3 - 4 | 535 | 75 | |
EU249757.1 | - | 3 - 4 | 555 | 75 |