Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148897.2 | NP_683695.1 | 1 - 2 | 446 | 64 |
GenBank mRNA | AK122782.1 | - | 1 - 2 | 446 | 64 |
AY044434.1 | - | 1 - 2 | 302 | 64 | |
BC101551.1 | - | 1 - 2 | 342 | 64 | |
BC101553.1 | - | 1 - 2 | 342 | 64 |