Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144972.1 | NP_001138444.1 | 1 - 2 | 161 | 59 |
NM_153337.2 | NP_699168.1 | 1 - 2 | 161 | 59 | |
NM_182854.3 | NP_878274.1 | 1 - 2 | 161 | 59 | |
GenBank mRNA | AA278249.1 | - | 1 - 2 | 162 | 59 |
AK055837.1 | - | 1 - 2 | 126 | 59 | |
AK292650.1 | - | 1 - 2 | 120 | 59 | |
AK292705.1 | - | 1 - 2 | 126 | 59 | |
AY302441.1 | - | 1 - 2 | 63 | 59 | |
BC027944.1 | - | 1 - 2 | 110 | 59 | |
BC063423.1 | - | 1 - 2 | 102 | 59 |