Assay Design



Gene Transcripts

Gene Symbol
MEIS2
Entrez Gene ID
4212
Gene Name
Meis homeobox 2
Gene Aliases
HsT18361, MRG1
Chromosome Location
Chr.15: 36891021 - 37101299 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002399.3 NP_002390.1 12 - 13 1625 74
NM_170674.4 NP_733774.1 12 - 13 1903 74
NM_170677.4 NP_733777.1 12 - 13 1924 74
NM_172315.2 NP_758526.1 11 - 12 1400 74
NM_172316.2 NP_758527.1 12 - 13 1690 74
NR_051953.1 11 - 12 1826 74
XM_006720524.1 XP_006720587.1 12 - 13 1314 74
XM_006720525.1 XP_006720588.1 12 - 13 1293 74
XM_006720529.2 XP_006720592.1 11 - 12 2191 74
XM_017022205.1 XP_016877694.1 12 - 13 2289 74
GenBank mRNA AF017418.1 - 5 - 6 420 73
AF178948.1 - 12 - 13 1505 74
AF179896.1 - 12 - 13 1484 74
AF179899.1 - 11 - 12 1407 74
AK055936.1 - 11 - 12 1124 74
AK056038.1 - 11 - 12 1341 74
AK056216.1 - 12 - 13 2323 74
AK056620.1 - 12 - 13 1678 74
AK226072.1 - 12 - 13 1927 74
AK294411.1 - 10 - 11 1078 74
BC001516.2 - 12 - 13 1629 74
BC001844.2 - 12 - 13 1629 74
BC007202.1 - 12 - 13 1629 74

View Full Product Details