Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173039.2 | NP_766627.1 | 1 - 2 | 975 | 80 |
GenBank mRNA | AB028147.1 | - | 1 - 2 | 922 | 80 |
AK075346.1 | - | 1 - 2 | 617 | 80 | |
AK311096.1 | - | 2 - 3 | 372 | 80 | |
BC040443.1 | - | 1 - 2 | 975 | 80 |