Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033576.1 | NP_001028748.1 | 15 - 16 | 2121 | 65 |
NM_001267052.1 | NP_001253981.1 | 15 - 16 | 2118 | 65 | |
NM_001308281.1 | NP_001295210.1 | 12 - 13 | 1787 | 65 | |
NM_173167.3 | NP_775259.1 | 14 - 15 | 2030 | 65 | |
XM_017024234.1 | XP_016879723.1 | 13 - 14 | 1905 | 65 | |
GenBank mRNA | AF539794.1 | - | 14 - 15 | 2030 | 65 |
AL832355.1 | - | 15 - 16 | 2115 | 65 | |
AL833281.1 | - | 15 - 16 | 2094 | 65 | |
BC101062.2 | - | 12 - 13 | 1787 | 65 | |
BC101063.2 | - | 14 - 15 | 2024 | 65 | |
BX647642.1 | - | 15 - 16 | 2121 | 65 |