Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004416.2 | NP_001004416.2 | 20 - 21 | 3684 | 73 |
NM_001199527.1 | NP_001186456.1 | 19 - 20 | 4248 | 73 | |
NM_001199528.2 | NP_001186457.2 | 20 - 21 | 3864 | 73 | |
NM_173568.3 | NP_775839.3 | 19 - 20 | 4068 | 73 | |
XM_011529797.1 | XP_011528099.1 | 17 - 18 | 3676 | 73 | |
XM_017028506.1 | XP_016883995.1 | 19 - 20 | 4063 | 73 | |
XM_017028507.1 | XP_016883996.1 | 19 - 20 | 6992 | 73 | |
XM_017028508.1 | XP_016883997.1 | 9 - 10 | 1847 | 73 | |
GenBank mRNA | AB051810.1 | - | 19 - 20 | 4248 | 73 |
AB051811.1 | - | 20 - 21 | 3864 | 73 | |
AK093654.1 | - | 5 - 6 | 926 | 73 | |
AK124223.1 | - | 6 - 7 | 1428 | 73 | |
AY771619.1 | - | 19 - 20 | 4068 | 73 | |
AY771620.1 | - | 20 - 21 | 3684 | 73 |