Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130918.1 | NP_001124390.1 | 14 - 15 | 2449 | 93 |
NM_173623.3 | NP_775894.2 | 7 - 8 | 1537 | 93 | |
XM_017024491.1 | XP_016879980.1 | 14 - 15 | 2592 | 93 | |
XM_017024492.1 | XP_016879981.1 | 13 - 14 | 2705 | 93 | |
XM_017024493.1 | XP_016879982.1 | 15 - 16 | 2659 | 93 | |
XM_017024494.1 | XP_016879983.1 | 14 - 15 | 2594 | 93 | |
XM_017024495.1 | XP_016879984.1 | 13 - 14 | 3322 | 93 | |
XM_017024496.1 | XP_016879985.1 | 9 - 10 | 1732 | 93 | |
GenBank mRNA | AK093127.1 | - | 6 - 7 | 1401 | 93 |
AK302952.1 | - | 14 - 15 | 2395 | 93 | |
BC033563.1 | - | 7 - 8 | 1538 | 93 | |
BC041368.2 | - | 14 - 15 | 3445 | 93 |