Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173653.3 | NP_775924.1 | 6 - 7 | 954 | 82 |
XM_011512703.2 | XP_011511005.1 | 3 - 4 | 264 | 82 | |
XM_011512704.2 | XP_011511006.1 | 6 - 7 | 927 | 82 | |
XM_017006202.1 | XP_016861691.1 | 6 - 7 | 927 | 82 | |
XM_017006203.1 | XP_016861692.1 | 5 - 6 | 712 | 82 | |
GenBank mRNA | AB089794.1 | - | 6 - 7 | 745 | 82 |
AK301816.1 | - | 5 - 6 | 680 | 82 | |
AK315107.1 | - | 6 - 7 | 881 | 82 | |
AL832304.1 | - | 6 - 7 | 819 | 82 | |
AY254100.1 | - | 6 - 7 | 869 | 82 | |
BC035779.1 | - | 6 - 7 | 870 | 82 |