Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173653.3 | NP_775924.1 | 12 - 13 | 1682 | 109 |
XM_011512703.2 | XP_011511005.1 | 9 - 10 | 992 | 109 | |
XM_017006202.1 | XP_016861691.1 | 12 - 13 | 1655 | 109 | |
XM_017006203.1 | XP_016861692.1 | 11 - 12 | 1440 | 109 | |
GenBank mRNA | AB075486.1 | - | 2 - 3 | 183 | 109 |
AB089794.1 | - | 12 - 13 | 1473 | 109 | |
AK092932.1 | - | 6 - 7 | 733 | 109 | |
AK315107.1 | - | 12 - 13 | 1609 | 109 | |
AL832304.1 | - | 12 - 13 | 1547 | 109 | |
AY254100.1 | - | 12 - 13 | 1597 | 109 | |
BC035779.1 | - | 12 - 13 | 1598 | 109 |