Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001700.4 | NP_001691.1 | 4 - 5 | 620 | 125 |
GenBank mRNA | BC069495.1 | - | 4 - 5 | 615 | 125 |
BC093931.1 | - | 4 - 5 | 620 | 125 | |
BC093933.1 | - | 4 - 5 | 620 | 125 | |
X58794.1 | - | 4 - 5 | 593 | 125 |