Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001843.3 | NP_001834.2 | 17 - 18 | 2337 | 83 |
NM_175038.2 | NP_778203.1 | 15 - 16 | 2153 | 83 | |
XM_005268651.2 | XP_005268708.1 | 17 - 18 | 2700 | 83 | |
XM_006719241.2 | XP_006719304.1 | 17 - 18 | 2324 | 83 | |
XM_011537926.2 | XP_011536228.1 | 17 - 18 | 2289 | 83 | |
XM_011537927.2 | XP_011536229.1 | 17 - 18 | 2346 | 83 | |
XM_017018821.1 | XP_016874310.1 | 17 - 18 | 2340 | 83 | |
XM_017018822.1 | XP_016874311.1 | 17 - 18 | 2289 | 83 | |
XM_017018823.1 | XP_016874312.1 | 17 - 18 | 2700 | 83 | |
XM_017018824.1 | XP_016874313.1 | 17 - 18 | 2324 | 83 | |
GenBank mRNA | U07819.1 | - | 16 - 17 | 2110 | 83 |
U07820.1 | - | 15 - 16 | 2158 | 83 | |
Z21488.1 | - | 17 - 18 | 2231 | 83 |