Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136025.4 | NP_001129497.1 | 2 - 3 | 208 | 120 |
NM_001172335.2 | NP_001165806.1 | 2 - 3 | 208 | 120 | |
NM_005032.6 | NP_005023.2 | 2 - 3 | 493 | 120 | |
XM_011537534.1 | XP_011535836.1 | 2 - 3 | 184 | 120 | |
GenBank mRNA | AK298501.1 | - | 2 - 3 | 169 | 120 |
AK298530.1 | - | 2 - 3 | 196 | 120 | |
AK311211.1 | - | 2 - 3 | 149 | 120 | |
AK312391.1 | - | 2 - 3 | 167 | 120 | |
BC039049.1 | - | 2 - 3 | 107 | 120 | |
BC056898.1 | - | 2 - 3 | 211 | 120 | |
BP214207.1 | - | 2 - 3 | 207 | 120 | |
DC320695.1 | - | 2 - 3 | 493 | 120 | |
DC357886.1 | - | 2 - 3 | 208 | 120 | |
DC416446.1 | - | 2 - 3 | 464 | 120 | |
M34427.1 | - | 2 - 3 | 161 | 120 |