Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136025.4 | NP_001129497.1 | 7 - 8 | 883 | 73 |
NM_001172335.2 | NP_001165806.1 | 7 - 8 | 802 | 73 | |
NM_001282337.1 | NP_001269266.1 | 8 - 9 | 1287 | 73 | |
NM_001282338.1 | NP_001269267.1 | 8 - 9 | 1265 | 73 | |
NM_005032.6 | NP_005023.2 | 7 - 8 | 1168 | 73 | |
XM_011537534.1 | XP_011535836.1 | 7 - 8 | 859 | 73 | |
GenBank mRNA | AK294509.1 | - | 8 - 9 | 909 | 73 |
AK298530.1 | - | 7 - 8 | 790 | 73 | |
AK300575.1 | - | 8 - 9 | 1219 | 73 | |
AK312391.1 | - | 7 - 8 | 842 | 73 | |
BC008588.1 | - | 1 - 2 | 88 | 73 | |
BC039049.1 | - | 7 - 8 | 782 | 73 | |
BC056898.1 | - | 7 - 8 | 886 | 73 | |
M22299.1 | - | 5 - 6 | 569 | 73 |