Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005076.3 | NP_005067.1 | 11 - 12 | 1662 | 61 |
XM_017002198.1 | XP_016857687.1 | 10 - 11 | 1403 | 61 | |
XM_017002199.1 | XP_016857688.1 | 12 - 13 | 1917 | 61 | |
GenBank mRNA | AK124736.1 | - | 6 - 7 | 1385 | 61 |
AK289985.1 | - | 10 - 11 | 1403 | 61 | |
BC129986.1 | - | 10 - 11 | 1428 | 61 | |
CR749495.1 | - | 10 - 11 | 2509 | 61 | |
X67734.1 | - | 11 - 12 | 1574 | 61 | |
X68274.1 | - | 11 - 12 | 1568 | 61 |