Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005173.3 | NP_005164.2 | 15 - 16 | 2498 | 67 |
NM_174953.2 | NP_777613.1 | 15 - 16 | 2498 | 67 | |
NM_174954.2 | NP_777614.1 | 15 - 16 | 2498 | 67 | |
NM_174955.2 | NP_777615.1 | 15 - 16 | 2498 | 67 | |
NM_174956.2 | NP_777616.1 | 15 - 16 | 2498 | 67 | |
NM_174957.2 | NP_777617.1 | 15 - 16 | 2498 | 67 | |
NM_174958.2 | NP_777618.1 | 15 - 16 | 2498 | 67 | |
XM_011523881.2 | XP_011522183.1 | 15 - 16 | 2498 | 67 | |
XM_011523882.2 | XP_011522184.1 | 15 - 16 | 2498 | 67 | |
XM_011523884.2 | XP_011522186.1 | 15 - 16 | 2498 | 67 | |
XM_011523885.1 | XP_011522187.1 | 15 - 16 | 2498 | 67 | |
XM_011523888.2 | XP_011522190.1 | 15 - 16 | 2498 | 67 | |
XM_011523889.1 | XP_011522191.1 | 15 - 16 | 2498 | 67 | |
XM_017024692.1 | XP_016880181.1 | 15 - 16 | 2498 | 67 | |
XM_017024693.1 | XP_016880182.1 | 13 - 14 | 2124 | 67 | |
GenBank mRNA | AF068220.1 | - | 15 - 16 | 2449 | 67 |
AF068221.1 | - | 3 - 4 | 726 | 67 | |
AK292716.1 | - | 15 - 16 | 2498 | 67 | |
BC035729.1 | - | 15 - 16 | 2453 | 67 | |
Z69881.1 | - | 15 - 16 | 2331 | 67 |