Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145513.1 | NP_001138985.1 | 6 - 7 | 1084 | 88 |
NM_001145514.1 | NP_001138986.1 | 6 - 7 | 1016 | 88 | |
NM_001145515.1 | NP_001138987.1 | 5 - 6 | 916 | 88 | |
NM_014766.4 | NP_055581.3 | 6 - 7 | 1076 | 88 | |
GenBank mRNA | AB071705.1 | - | 6 - 7 | 966 | 88 |
AK289514.1 | - | 5 - 6 | 1111 | 88 | |
AK294891.1 | - | 3 - 4 | 446 | 88 | |
AK295172.1 | - | 6 - 7 | 1016 | 88 | |
AK295713.1 | - | 5 - 6 | 819 | 88 | |
AK295800.1 | - | 3 - 4 | 435 | 88 | |
BC040492.1 | - | 6 - 7 | 1028 | 88 | |
D83777.2 | - | 6 - 7 | 1021 | 88 |