Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_078471.3 | NP_510880.2 | 26 - 27 | 4267 | 77 |
NM_203318.1 | NP_976063.1 | 26 - 27 | 4267 | 77 | |
GenBank mRNA | AB177858.1 | - | 25 - 26 | 4086 | 77 |
AB177860.1 | - | 26 - 27 | 3219 | 77 | |
AK302505.1 | - | 25 - 26 | 3020 | 77 | |
AK302725.1 | - | 10 - 11 | 981 | 77 | |
AK303522.1 | - | 25 - 26 | 3157 | 77 | |
AY703984.1 | - | 26 - 27 | 3219 | 77 | |
BC039612.1 | - | 26 - 27 | 4371 | 77 | |
CR933614.1 | - | 26 - 27 | 4242 | 77 | |
D86970.2 | - | 26 - 27 | 4290 | 77 |