Assay Design



Gene Transcripts

Gene Symbol
MYO18A
Entrez Gene ID
399687
Gene Name
myosin XVIIIA
Gene Aliases
MYSPDZ, SPR210
Chromosome Location
Chr.17: 29073510 - 29180389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078471.3 NP_510880.2 34 - 35 5342 79
NM_203318.1 NP_976063.1 34 - 35 5342 79
GenBank mRNA AB177858.1 - 33 - 34 5161 79
AB177860.1 - 34 - 35 4294 79
AK302505.1 - 33 - 34 4068 79
AK302725.1 - 18 - 19 2056 79
AY703984.1 - 34 - 35 4294 79
BC039612.1 - 33 - 34 5335 79
CR933614.1 - 34 - 35 5317 79
D86970.2 - 34 - 35 5365 79

View Full Product Details