Assay Design
Gene Transcripts
- Gene Symbol
- EYA3
- Entrez Gene ID
-
2140
- Gene Name
- EYA transcriptional coactivator and phosphatase 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 27970344 - 28088671 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_104214.1 |
|
13 - 14 |
1338 |
87 |
GenBank mRNA |
AK303664.1 |
- |
11 - 12 |
1114 |
87 |
BC029500.1 |
- |
13 - 14 |
1298 |
87 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI743819.1 |
- |
4 - 5 |
|
|
Chr. 1 - 27988477 - 28000051 [+] on Build GRCh38 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
XK related 8 |
XKR8 |
XRG8, hXkr8 |
-29.2kb
|
Anti-sense |
Chr. - 27959462 - 27968096 on Build GRCh38 |
Downstream |
platelet activating factor receptor |
PTAFR |
PAFR |
148.8kb
|
Sense |
Chr. - 28147166 - 28193936 on Build GRCh38 |
View Full Product Details