Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000070.2 | NP_000061.1 | 18 - 19 | 2359 | 66 |
NM_024344.1 | NP_077320.1 | 17 - 18 | 2341 | 66 | |
NM_173087.1 | NP_775110.1 | 15 - 16 | 2083 | 66 | |
NM_173088.1 | NP_775111.1 | 7 - 8 | 686 | 66 | |
NM_173089.1 | NP_775112.1 | 3 - 4 | 316 | 66 | |
NM_173090.1 | NP_775113.1 | 4 - 5 | 334 | 66 | |
GenBank mRNA | AF127764.1 | - | 17 - 18 | 2256 | 66 |
AF127765.3 | - | 15 - 16 | 1998 | 66 | |
AK027249.1 | - | 2 - 3 | 976 | 66 | |
BC003169.1 | - | 7 - 8 | 686 | 66 | |
BC003521.1 | - | 3 - 4 | 298 | 66 | |
BC004883.2 | - | 3 - 4 | 316 | 66 | |
BC007810.2 | - | 4 - 5 | 282 | 66 | |
BC067126.1 | - | 3 - 4 | 299 | 66 | |
BC100782.1 | - | 3 - 4 | 297 | 66 | |
BC100783.1 | - | 3 - 4 | 297 | 66 | |
BC100784.2 | - | 3 - 4 | 297 | 66 | |
BC107791.1 | - | 4 - 5 | 449 | 66 | |
BC128605.1 | - | 3 - 4 | 297 | 66 | |
BC146649.1 | - | 18 - 19 | 2272 | 66 | |
BC146672.1 | - | 18 - 19 | 2272 | 66 | |
BT007322.1 | - | 6 - 7 | 517 | 66 | |
EU791850.1 | - | 18 - 19 | 1792 | 66 | |
EU791851.1 | - | 17 - 18 | 1648 | 66 | |
KU177954.1 | - | 6 - 7 | 517 | 66 | |
KU177955.1 | - | 6 - 7 | 618 | 66 | |
KU177956.1 | - | 4 - 5 | 385 | 66 | |
X85030.1 | - | 18 - 19 | 2053 | 66 |