Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102948956-102949229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP13
Entrez Gene ID
4322
Gene Name
matrix metallopeptidase 13
Gene Aliases
CLG3, MANDP1, MMP-13
UniGene
Hs.2936
Gene Chromosome Location
Chr.11: 102942992-102955734 on Build GRCh38
Transcripts
NM_002427.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368185 c.1015G>A p.E339K Chr.11: 102949061-102949061 on Build GRCh38
1603956 c.918-2A>G p.? Chr.11: 102949160-102949160 on Build GRCh38
6713747 c.928C>T p.R310C Chr.11: 102949148-102949148 on Build GRCh38
6713746 c.1028A>G p.H343R Chr.11: 102949048-102949048 on Build GRCh38
1746008 c.1041C>A p.F347L Chr.11: 102949035-102949035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782354118 Chr.11: 102949179-102949179 on Build GRCh38
rs759581047 Chr.11: 102949122-102949122 on Build GRCh38
rs185832993 Chr.11: 102949108-102949108 on Build GRCh38
rs376188703 Chr.11: 102949013-102949013 on Build GRCh38
rs368129116 Chr.11: 102949192-102949192 on Build GRCh38
rs782152104 Chr.11: 102949079-102949079 on Build GRCh38
rs782252957 Chr.11: 102949188-102949188 on Build GRCh38
rs781880258 Chr.11: 102949127-102949127 on Build GRCh38
rs782060589 Chr.11: 102948990-102948990 on Build GRCh38
rs782093889 Chr.11: 102949117-102949117 on Build GRCh38
rs371762358 Chr.11: 102949146-102949146 on Build GRCh38
rs782588636 Chr.11: 102949164-102949164 on Build GRCh38
rs782597212 Chr.11: 102949107-102949107 on Build GRCh38
rs142601143 Chr.11: 102949141-102949141 on Build GRCh38
rs782035543 Chr.11: 102949078-102949078 on Build GRCh38
rs782445474 Chr.11: 102949140-102949140 on Build GRCh38
rs781834730 Chr.11: 102948998-102948998 on Build GRCh38
rs782718133 Chr.11: 102949119-102949119 on Build GRCh38
rs782377946 Chr.11: 102949076-102949076 on Build GRCh38
rs782536757 Chr.11: 102949159-102949159 on Build GRCh38
rs782185609 Chr.11: 102949022-102949022 on Build GRCh38
rs201001079 Chr.11: 102949194-102949194 on Build GRCh38
rs78356340 Chr.11: 102949187-102949187 on Build GRCh38
rs782358497 Chr.11: 102949034-102949034 on Build GRCh38
rs201043020 Chr.11: 102949174-102949174 on Build GRCh38
rs782535983 Chr.11: 102949147-102949147 on Build GRCh38
rs782582945 Chr.11: 102949014-102949014 on Build GRCh38
rs782615439 Chr.11: 102949071-102949071 on Build GRCh38
rs367612153 Chr.11: 102949125-102949125 on Build GRCh38
rs375347249 Chr.11: 102949012-102949012 on Build GRCh38
rs199992289 Chr.11: 102949123-102949123 on Build GRCh38
rs542014947 Chr.11: 102949000-102949000 on Build GRCh38
rs782703007 Chr.11: 102948996-102948996 on Build GRCh38
rs782298324 Chr.11: 102949149-102949149 on Build GRCh38
rs781847523 Chr.11: 102949010-102949010 on Build GRCh38
rs782180276 Chr.11: 102949170-102949170 on Build GRCh38
rs375641186 Chr.11: 102948987-102948987 on Build GRCh38
rs782628284 Chr.11: 102949148-102949148 on Build GRCh38
rs370752191 Chr.11: 102949007-102949007 on Build GRCh38

View Full Product Details