Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52315169-52315432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940671 c.1293C>A p.V431V Chr.12: 52315313-52315313 on Build GRCh38
5779673 c.1285G>T p.V429L Chr.12: 52315321-52315321 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748320119 Chr.12: 52315386-52315386 on Build GRCh38
rs756804256 Chr.12: 52315340-52315340 on Build GRCh38
rs984930937 Chr.12: 52315207-52315207 on Build GRCh38
rs371725329 Chr.12: 52315302-52315302 on Build GRCh38
rs374805734 Chr.12: 52315387-52315387 on Build GRCh38
rs774501121 Chr.12: 52315276-52315276 on Build GRCh38
rs913037813 Chr.12: 52315251-52315251 on Build GRCh38
rs898765329 Chr.12: 52315370-52315370 on Build GRCh38
rs990004031 Chr.12: 52315250-52315250 on Build GRCh38
rs773205109 Chr.12: 52315290-52315290 on Build GRCh38
rs781156149 Chr.12: 52315265-52315265 on Build GRCh38
rs752305742 Chr.12: 52315330-52315330 on Build GRCh38
rs978736201 Chr.12: 52315346-52315346 on Build GRCh38
rs745811112 Chr.12: 52315351-52315351 on Build GRCh38
rs1034169997 Chr.12: 52315212-52315212 on Build GRCh38
rs779657085 Chr.12: 52315305-52315308 on Build GRCh38
rs779837406 Chr.12: 52315364-52315364 on Build GRCh38
rs772148240 Chr.12: 52315285-52315285 on Build GRCh38
rs762078992 Chr.12: 52315280-52315280 on Build GRCh38
rs112868674 Chr.12: 52315299-52315299 on Build GRCh38
rs754341942 Chr.12: 52315304-52315304 on Build GRCh38
rs377257286 Chr.12: 52315399-52315399 on Build GRCh38
rs369869028 Chr.12: 52315407-52315407 on Build GRCh38
rs980448697 Chr.12: 52315203-52315203 on Build GRCh38
rs148757217 Chr.12: 52315338-52315338 on Build GRCh38
rs191425339 Chr.12: 52315320-52315320 on Build GRCh38
rs769532480 Chr.12: 52315358-52315358 on Build GRCh38
rs1040082297 Chr.12: 52315367-52315367 on Build GRCh38
rs758002752 Chr.12: 52315337-52315337 on Build GRCh38
rs756224896 Chr.12: 52315211-52315211 on Build GRCh38
rs3825221 Chr.12: 52315194-52315194 on Build GRCh38
rs746526075 Chr.12: 52315393-52315423 on Build GRCh38
rs754111437 Chr.12: 52315307-52315307 on Build GRCh38
rs769599188 Chr.12: 52315271-52315271 on Build GRCh38
rs773405486 Chr.12: 52315391-52315391 on Build GRCh38
rs770997363 Chr.12: 52315400-52315400 on Build GRCh38
rs918062198 Chr.12: 52315310-52315313 on Build GRCh38
rs3741721 Chr.12: 52315248-52315248 on Build GRCh38
rs772237832 Chr.12: 52315389-52315389 on Build GRCh38
rs781317580 Chr.12: 52315343-52315343 on Build GRCh38
rs760786648 Chr.12: 52315393-52315393 on Build GRCh38
rs760032840 Chr.12: 52315408-52315408 on Build GRCh38
rs745698011 Chr.12: 52315266-52315266 on Build GRCh38
rs935895322 Chr.12: 52315249-52315249 on Build GRCh38

View Full Product Details