Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323011.1 | NP_001309940.1 | 2 - 3 | 136 | 114 |
NM_001323012.1 | NP_001309941.1 | 2 - 3 | 143 | 114 | |
NM_006556.3 | NP_006547.1 | 2 - 3 | 486 | 114 | |
GenBank mRNA | AK130234.1 | - | 3 - 4 | 328 | 114 |
AK312803.1 | - | 2 - 3 | 445 | 114 | |
BC006089.1 | - | 2 - 3 | 469 | 114 | |
BC007694.2 | - | 2 - 3 | 399 | 114 | |
BQ061044.1 | - | 2 - 3 | 486 | 114 | |
BT019976.1 | - | 2 - 3 | 163 | 114 | |
CR541998.1 | - | 2 - 3 | 163 | 114 | |
L77213.1 | - | 2 - 3 | 198 | 114 |