Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 10 - 11 | 1677 | 91 |
GenBank mRNA | AF029232.1 | - | 10 - 11 | 1577 | 91 |
AJ000388.1 | - | 10 - 11 | 1668 | 91 | |
AK300340.1 | - | 8 - 9 | 1233 | 91 | |
AK312701.1 | - | 10 - 11 | 1641 | 91 | |
BC000730.2 | - | 9 - 10 | 1506 | 91 |