Assay Design



Gene Transcripts

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
-
Chromosome Location
Chr.17: 68259089 - 68452000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267727.1 NP_001254656.1 2 - 3 841 69
NM_014960.4 NP_055775.2 2 - 3 1012 69
XM_005257170.3 XP_005257227.1 2 - 3 1025 69
XM_005257172.3 XP_005257229.1 2 - 3 1074 69
XM_006721777.3 XP_006721840.2 2 - 3 1039 69
XM_006721779.3 XP_006721842.1 2 - 3 1066 69
XM_011524535.2 XP_011522837.1 2 - 3 1037 69
XM_011524536.2 XP_011522838.1 2 - 3 867 69
XM_011524537.1 XP_011522839.1 2 - 3 788 69
XM_011524538.2 XP_011522840.1 2 - 3 1039 69
XM_011524540.2 XP_011522842.1 2 - 3 1062 69
XM_011524541.2 XP_011522843.1 2 - 3 1105 69
XM_011524542.2 XP_011522844.1 2 - 3 1105 69
XM_011524543.2 XP_011522845.1 2 - 3 1105 69
XM_011524544.2 XP_011522846.1 2 - 3 1105 69
XM_011524545.2 XP_011522847.1 2 - 3 1063 69
XM_017024360.1 XP_016879849.1 2 - 3 877 69
XM_017024361.1 XP_016879850.1 2 - 3 1025 69
XM_017024362.1 XP_016879851.1 2 - 3 964 69
XM_017024363.1 XP_016879852.1 2 - 3 870 69
XM_017024364.1 XP_016879853.1 2 - 3 866 69
XM_017024365.1 XP_016879854.1 2 - 3 854 69
XM_017024366.1 XP_016879855.1 2 - 3 1065 69
XM_017024367.1 XP_016879856.1 2 - 3 867 69
XM_017024368.1 XP_016879857.1 2 - 3 867 69
GenBank mRNA AB023218.1 - 1 - 2 674 69
AY358380.1 - 2 - 3 1012 69
BC012375.1 - 1 - 2 255 69

View Full Product Details