Assay Design
Gene Transcripts
- Gene Symbol
- HIST1H2AD
- Entrez Gene ID
-
3013
- Gene Name
- histone cluster 1, H2ad
- Gene Aliases
-
H2A.3, H2A/g, H2AFG
- Chromosome Location
-
Chr.6: 26198784 - 26199293 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_021065.3 |
NP_066409.1 |
1 - 1 |
440 |
97 |
GenBank mRNA |
BC128035.1 |
- |
1 - 1 |
390 |
97 |
View Full Product Details