Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67231345-67231614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHOD1
Entrez Gene ID
29109
Gene Name
formin homology 2 domain containing 1
Gene Aliases
FHOS
UniGene
Hs.95231
Gene Chromosome Location
Chr.16: 67229389-67247522 on Build GRCh38
Transcripts
NM_001318202.1 NM_013241.2 XM_006721180.1 XM_011523043.2 XM_011523044.1 XM_011523045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4971156 c.2465C>T p.A822V Chr.16: 67231470-67231470 on Build GRCh38
5619117 c.2478G>T p.A826A Chr.16: 67231457-67231457 on Build GRCh38
6036531 c.2478G>A p.A826A Chr.16: 67231457-67231457 on Build GRCh38
6522930 c.2434G>T p.V812L Chr.16: 67231501-67231501 on Build GRCh38
6663968 c.2453G>A p.R818H Chr.16: 67231482-67231482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376122137 Chr.16: 67231528-67231528 on Build GRCh38
rs200196443 Chr.16: 67231402-67231402 on Build GRCh38
rs767135405 Chr.16: 67231465-67231465 on Build GRCh38
rs750037896 Chr.16: 67231479-67231479 on Build GRCh38
rs937031607 Chr.16: 67231392-67231392 on Build GRCh38
rs754121800 Chr.16: 67231493-67231493 on Build GRCh38
rs768452969 Chr.16: 67231451-67231451 on Build GRCh38
rs745657573 Chr.16: 67231424-67231424 on Build GRCh38
rs963016718 Chr.16: 67231585-67231585 on Build GRCh38
rs143478109 Chr.16: 67231529-67231529 on Build GRCh38
rs760414850 Chr.16: 67231582-67231582 on Build GRCh38
rs747674590 Chr.16: 67231565-67231565 on Build GRCh38
rs758202378 Chr.16: 67231574-67231575 on Build GRCh38
rs771659014 Chr.16: 67231567-67231567 on Build GRCh38
rs765739030 Chr.16: 67231589-67231589 on Build GRCh38
rs779006370 Chr.16: 67231390-67231390 on Build GRCh38
rs755559629 Chr.16: 67231505-67231505 on Build GRCh38
rs778551876 Chr.16: 67231421-67231424 on Build GRCh38
rs753148833 Chr.16: 67231592-67231592 on Build GRCh38
rs749021162 Chr.16: 67231538-67231538 on Build GRCh38
rs76116994 Chr.16: 67231399-67231399 on Build GRCh38
rs779379018 Chr.16: 67231510-67231510 on Build GRCh38
rs769576846 Chr.16: 67231430-67231430 on Build GRCh38
rs752842034 Chr.16: 67231518-67231518 on Build GRCh38
rs779945795 Chr.16: 67231537-67231537 on Build GRCh38
rs766724851 Chr.16: 67231484-67231484 on Build GRCh38
rs772340071 Chr.16: 67231403-67231403 on Build GRCh38
rs745866759 Chr.16: 67231465-67231465 on Build GRCh38
rs777114291 Chr.16: 67231583-67231583 on Build GRCh38
rs542514076 Chr.16: 67231456-67231456 on Build GRCh38
rs909003517 Chr.16: 67231422-67231422 on Build GRCh38
rs755042411 Chr.16: 67231380-67231380 on Build GRCh38
rs774360642 Chr.16: 67231551-67231551 on Build GRCh38
rs772768944 Chr.16: 67231570-67231570 on Build GRCh38
rs777971404 Chr.16: 67231413-67231413 on Build GRCh38
rs759814824 Chr.16: 67231586-67231586 on Build GRCh38
rs762986161 Chr.16: 67231435-67231435 on Build GRCh38
rs768776568 Chr.16: 67231541-67231541 on Build GRCh38
rs199498142 Chr.16: 67231491-67231491 on Build GRCh38
rs772699637 Chr.16: 67231534-67231534 on Build GRCh38
rs754321216 Chr.16: 67231376-67231376 on Build GRCh38
rs200521596 Chr.16: 67231433-67231433 on Build GRCh38
rs6499118 Chr.16: 67231457-67231457 on Build GRCh38
rs145700198 Chr.16: 67231483-67231483 on Build GRCh38
rs66985814 Chr.16: 67231414-67231420 on Build GRCh38

View Full Product Details