Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 13562675-13562912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIN2B
Entrez Gene ID
2904
Gene Name
glutamate ionotropic receptor NMDA type subunit 2B
Gene Aliases
EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
UniGene
Hs.504844
Gene Chromosome Location
Chr.12: 13537337-13982012 on Build GRCh38
Transcripts
NM_000834.3 XM_005253351.3 XM_011520628.2 XM_011520629.2 XM_017019219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575252 c.4395C>A p.F1465L Chr.12: 13562843-13562843 on Build GRCh38
1665489 c.4438_4439delAT p.I1480fs*1 Chr.12: 13562799-13562800 on Build GRCh38
3458451 c.4347G>A p.G1449G Chr.12: 13562891-13562891 on Build GRCh38
3368733 c.4423G>A p.E1475K Chr.12: 13562815-13562815 on Build GRCh38
937453 c.4382A>G p.N1461S Chr.12: 13562856-13562856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570811791 Chr.12: 13562725-13562725 on Build GRCh38
rs372520149 Chr.12: 13562769-13562769 on Build GRCh38
rs945611220 Chr.12: 13562719-13562719 on Build GRCh38
rs369455429 Chr.12: 13562858-13562858 on Build GRCh38
rs762140684 Chr.12: 13562789-13562789 on Build GRCh38
rs760660431 Chr.12: 13562750-13562750 on Build GRCh38
rs757954618 Chr.12: 13562859-13562859 on Build GRCh38
rs771877727 Chr.12: 13562732-13562732 on Build GRCh38
rs975747936 Chr.12: 13562699-13562699 on Build GRCh38
rs780659331 Chr.12: 13562854-13562854 on Build GRCh38
rs202051424 Chr.12: 13562825-13562825 on Build GRCh38
rs200127692 Chr.12: 13562838-13562838 on Build GRCh38
rs753929690 Chr.12: 13562767-13562768 on Build GRCh38
rs201145829 Chr.12: 13562829-13562829 on Build GRCh38
rs200778827 Chr.12: 13562720-13562720 on Build GRCh38
rs764339423 Chr.12: 13562782-13562782 on Build GRCh38
rs765505393 Chr.12: 13562723-13562723 on Build GRCh38
rs200052879 Chr.12: 13562758-13562758 on Build GRCh38
rs897186462 Chr.12: 13562761-13562761 on Build GRCh38
rs199872486 Chr.12: 13562722-13562722 on Build GRCh38
rs756790727 Chr.12: 13562883-13562883 on Build GRCh38
rs746663430 Chr.12: 13562881-13562881 on Build GRCh38
rs752290736 Chr.12: 13562855-13562855 on Build GRCh38
rs577649736 Chr.12: 13562865-13562865 on Build GRCh38
rs997974723 Chr.12: 13562703-13562703 on Build GRCh38
rs746822162 Chr.12: 13562747-13562747 on Build GRCh38
rs766688154 Chr.12: 13562771-13562771 on Build GRCh38
rs751758015 Chr.12: 13562784-13562784 on Build GRCh38
rs759518484 Chr.12: 13562753-13562753 on Build GRCh38
rs751398824 Chr.12: 13562755-13562758 on Build GRCh38
rs1011159439 Chr.12: 13562714-13562714 on Build GRCh38
rs777620880 Chr.12: 13562876-13562876 on Build GRCh38
rs202133231 Chr.12: 13562832-13562832 on Build GRCh38
rs887589398 Chr.12: 13562879-13562879 on Build GRCh38
rs183243118 Chr.12: 13562724-13562724 on Build GRCh38
rs200503380 Chr.12: 13562729-13562729 on Build GRCh38
rs76294779 Chr.12: 13562861-13562861 on Build GRCh38
rs201438415 Chr.12: 13562704-13562704 on Build GRCh38
rs900906898 Chr.12: 13562699-13562699 on Build GRCh38
rs537983876 Chr.12: 13562731-13562731 on Build GRCh38
rs201744355 Chr.12: 13562721-13562721 on Build GRCh38
rs769557169 Chr.12: 13562756-13562756 on Build GRCh38
rs201467236 Chr.12: 13562730-13562730 on Build GRCh38
rs756658425 Chr.12: 13562834-13562834 on Build GRCh38
rs1022182345 Chr.12: 13562717-13562717 on Build GRCh38
rs199819153 Chr.12: 13562828-13562828 on Build GRCh38
rs745597964 Chr.12: 13562887-13562887 on Build GRCh38
rs201107419 Chr.12: 13562777-13562777 on Build GRCh38
rs910793751 Chr.12: 13562806-13562806 on Build GRCh38
rs199528895 Chr.12: 13562713-13562713 on Build GRCh38
rs781050985 Chr.12: 13562885-13562885 on Build GRCh38
rs777531033 Chr.12: 13562742-13562742 on Build GRCh38
rs200820444 Chr.12: 13562752-13562752 on Build GRCh38

View Full Product Details