Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113415252-113415525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD2
Entrez Gene ID
1813
Gene Name
dopamine receptor D2
Gene Aliases
D2DR, D2R
UniGene
Hs.73893
Gene Chromosome Location
Chr.11: 113409595-113475279 on Build GRCh38
Transcripts
NM_000795.3 NM_016574.3 XM_017017296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443548 c.633G>A p.K211K Chr.11: 113415511-113415511 on Build GRCh38
923193 c.689G>A p.R230Q Chr.11: 113415455-113415455 on Build GRCh38
3443541 c.715C>T p.P239S Chr.11: 113415429-113415429 on Build GRCh38
1250699 c.680G>A p.R227H Chr.11: 113415464-113415464 on Build GRCh38
5802487 c.702_713del12 p.H235_A238delHLRA Chr.11: 113415431-113415442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550561280 Chr.11: 113415287-113415287 on Build GRCh38
rs746038059 Chr.11: 113415391-113415391 on Build GRCh38
rs748440860 Chr.11: 113415413-113415413 on Build GRCh38
rs199961951 Chr.11: 113415454-113415454 on Build GRCh38
rs758555712 Chr.11: 113415480-113415480 on Build GRCh38
rs549341605 Chr.11: 113415295-113415295 on Build GRCh38
rs920264425 Chr.11: 113415276-113415276 on Build GRCh38
rs1042823282 Chr.11: 113415334-113415334 on Build GRCh38
rs755559594 Chr.11: 113415486-113415486 on Build GRCh38
rs757536778 Chr.11: 113415390-113415390 on Build GRCh38
rs201628301 Chr.11: 113415433-113415433 on Build GRCh38
rs780041167 Chr.11: 113415397-113415397 on Build GRCh38
rs201954853 Chr.11: 113415400-113415400 on Build GRCh38
rs112204362 Chr.11: 113415409-113415409 on Build GRCh38
rs767153535 Chr.11: 113415445-113415445 on Build GRCh38
rs140638952 Chr.11: 113415481-113415481 on Build GRCh38
rs555646231 Chr.11: 113415377-113415377 on Build GRCh38
rs987305741 Chr.11: 113415489-113415489 on Build GRCh38
rs755950380 Chr.11: 113415375-113415375 on Build GRCh38
rs566751371 Chr.11: 113415322-113415322 on Build GRCh38
rs1044689260 Chr.11: 113415296-113415296 on Build GRCh38
rs151320768 Chr.11: 113415470-113415470 on Build GRCh38
rs759201066 Chr.11: 113415444-113415444 on Build GRCh38
rs750647475 Chr.11: 113415479-113415479 on Build GRCh38
rs770234535 Chr.11: 113415420-113415420 on Build GRCh38
rs747562843 Chr.11: 113415405-113415405 on Build GRCh38
rs904244813 Chr.11: 113415335-113415335 on Build GRCh38
rs200419139 Chr.11: 113415455-113415455 on Build GRCh38
rs1003073966 Chr.11: 113415312-113415312 on Build GRCh38
rs774178472 Chr.11: 113415424-113415424 on Build GRCh38
rs371023969 Chr.11: 113415464-113415464 on Build GRCh38
rs201881891 Chr.11: 113415465-113415465 on Build GRCh38
rs764968856 Chr.11: 113415477-113415477 on Build GRCh38
rs553233688 Chr.11: 113415320-113415320 on Build GRCh38
rs201316581 Chr.11: 113415403-113415404 on Build GRCh38
rs376532450 Chr.11: 113415437-113415437 on Build GRCh38
rs557036496 Chr.11: 113415485-113415485 on Build GRCh38
rs113260244 Chr.11: 113415324-113415324 on Build GRCh38
rs200148328 Chr.11: 113415488-113415488 on Build GRCh38
rs200527197 Chr.11: 113415395-113415395 on Build GRCh38
rs539782878 Chr.11: 113415309-113415309 on Build GRCh38
rs71653613 Chr.11: 113415349-113415349 on Build GRCh38
rs77989639 Chr.11: 113415286-113415286 on Build GRCh38
rs201004067 Chr.11: 113415427-113415427 on Build GRCh38
rs757062764 Chr.11: 113415467-113415467 on Build GRCh38
rs1017859677 Chr.11: 113415365-113415365 on Build GRCh38
rs769029308 Chr.11: 113415408-113415408 on Build GRCh38
rs906015051 Chr.11: 113415297-113415297 on Build GRCh38
rs201040625 Chr.11: 113415381-113415381 on Build GRCh38
rs752688406 Chr.11: 113415374-113415374 on Build GRCh38
rs200078397 Chr.11: 113415403-113415403 on Build GRCh38
rs781445866 Chr.11: 113415404-113415404 on Build GRCh38

View Full Product Details