Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22096864-22097135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_011545533.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs150619847 Chr.X: 22097023-22097023 on Build GRCh38
rs781343445 Chr.X: 22097090-22097090 on Build GRCh38
rs375559951 Chr.X: 22097053-22097053 on Build GRCh38
rs751961973 Chr.X: 22097078-22097078 on Build GRCh38
rs369652869 Chr.X: 22096904-22096904 on Build GRCh38
rs746380161 Chr.X: 22096940-22096940 on Build GRCh38
rs372663150 Chr.X: 22097027-22097027 on Build GRCh38
rs139771307 Chr.X: 22096972-22096972 on Build GRCh38
rs1055925604 Chr.X: 22096888-22096888 on Build GRCh38
rs201958235 Chr.X: 22097049-22097049 on Build GRCh38
rs776367122 Chr.X: 22096981-22096981 on Build GRCh38
rs754106853 Chr.X: 22097015-22097015 on Build GRCh38
rs58524357 Chr.X: 22097079-22097079 on Build GRCh38
rs746436151 Chr.X: 22096965-22096965 on Build GRCh38
rs772573976 Chr.X: 22096984-22096984 on Build GRCh38
rs767171186 Chr.X: 22097013-22097013 on Build GRCh38
rs759411768 Chr.X: 22097009-22097009 on Build GRCh38
rs769430757 Chr.X: 22096989-22096989 on Build GRCh38
rs780651981 Chr.X: 22097066-22097066 on Build GRCh38
rs950144148 Chr.X: 22096901-22096901 on Build GRCh38
rs753971660 Chr.X: 22096924-22096924 on Build GRCh38
rs193922460 Chr.X: 22096990-22096991 on Build GRCh38
rs886039582 Chr.X: 22096955-22096955 on Build GRCh38
rs762535711 Chr.X: 22097004-22097004 on Build GRCh38
rs768862054 Chr.X: 22096980-22096980 on Build GRCh38
rs367802921 Chr.X: 22096903-22096903 on Build GRCh38
rs778909953 Chr.X: 22096932-22096932 on Build GRCh38
rs866429868 Chr.X: 22096976-22096976 on Build GRCh38
rs1027914323 Chr.X: 22097060-22097060 on Build GRCh38
rs758973292 Chr.X: 22096943-22096943 on Build GRCh38
rs772921325 Chr.X: 22097000-22097000 on Build GRCh38
rs764420479 Chr.X: 22096914-22096914 on Build GRCh38
rs1017362124 Chr.X: 22096942-22096942 on Build GRCh38
rs747696209 Chr.X: 22096985-22096985 on Build GRCh38
rs142755818 Chr.X: 22097008-22097008 on Build GRCh38
rs754473894 Chr.X: 22096931-22096931 on Build GRCh38
rs767638519 Chr.X: 22096923-22096924 on Build GRCh38
rs1057517791 Chr.X: 22096988-22096988 on Build GRCh38
rs916000438 Chr.X: 22096900-22096900 on Build GRCh38
rs199893153 Chr.X: 22097006-22097006 on Build GRCh38
rs749088812 Chr.X: 22097101-22097101 on Build GRCh38

View Full Product Details