Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27919996-27920253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Gene Symbol
PDX1-AS1
Entrez Gene ID
100861550
Gene Name
PDX1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.416201
Gene Chromosome Location
Chr.13: 27829759-27921404 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4957641 c.27G>T p.A9A Chr.13: 27920165-27920165 on Build GRCh38
5970950 c.98C>A p.P33H Chr.13: 27920236-27920236 on Build GRCh38
4934793 c.97C>A p.P33T Chr.13: 27920235-27920235 on Build GRCh38
7383470 c.98C>T p.P33L Chr.13: 27920236-27920236 on Build GRCh38
1366181 c.16C>A p.Q6K Chr.13: 27920154-27920154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767400548 Chr.13: 27920089-27920102 on Build GRCh38
rs1015629646 Chr.13: 27920080-27920080 on Build GRCh38
rs557174897 Chr.13: 27920114-27920114 on Build GRCh38
rs913578374 Chr.13: 27920203-27920203 on Build GRCh38
rs146936598 Chr.13: 27920144-27920144 on Build GRCh38
rs776516205 Chr.13: 27920198-27920198 on Build GRCh38
rs759692004 Chr.13: 27920223-27920223 on Build GRCh38
rs867618159 Chr.13: 27920032-27920032 on Build GRCh38
rs146021107 Chr.13: 27920030-27920030 on Build GRCh38
rs1042602874 Chr.13: 27920057-27920057 on Build GRCh38
rs779510037 Chr.13: 27920115-27920115 on Build GRCh38
rs952795775 Chr.13: 27920073-27920073 on Build GRCh38
rs193922352 Chr.13: 27920116-27920117 on Build GRCh38
rs1024878476 Chr.13: 27920103-27920103 on Build GRCh38
rs954397231 Chr.13: 27920215-27920215 on Build GRCh38
rs1014037756 Chr.13: 27920096-27920096 on Build GRCh38
rs137852785 Chr.13: 27920190-27920190 on Build GRCh38
rs193922351 Chr.13: 27920121-27920121 on Build GRCh38
rs936474667 Chr.13: 27920166-27920166 on Build GRCh38
rs905029902 Chr.13: 27920043-27920043 on Build GRCh38
rs1000923448 Chr.13: 27920081-27920082 on Build GRCh38
rs188482874 Chr.13: 27920053-27920053 on Build GRCh38
rs1055620101 Chr.13: 27920082-27920082 on Build GRCh38
rs921718972 Chr.13: 27920209-27920209 on Build GRCh38
rs987574171 Chr.13: 27920216-27920216 on Build GRCh38
rs965979876 Chr.13: 27920081-27920081 on Build GRCh38
rs1025195080 Chr.13: 27920037-27920037 on Build GRCh38
rs773555653 Chr.13: 27920145-27920145 on Build GRCh38
rs201066590 Chr.13: 27920034-27920034 on Build GRCh38
rs192902098 Chr.13: 27920235-27920235 on Build GRCh38
rs747304604 Chr.13: 27920183-27920183 on Build GRCh38
rs767363575 Chr.13: 27920236-27920236 on Build GRCh38
rs978926784 Chr.13: 27920128-27920128 on Build GRCh38
rs1027085993 Chr.13: 27920070-27920070 on Build GRCh38
rs983184273 Chr.13: 27920074-27920074 on Build GRCh38
rs977032589 Chr.13: 27920100-27920100 on Build GRCh38
rs774060840 Chr.13: 27920089-27920095 on Build GRCh38

View Full Product Details