Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28399531-28399804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3515333 c.1368G>A p.Q456Q Chr.17: 28399665-28399665 on Build GRCh38
3819096 c.1344C>T p.H448H Chr.17: 28399689-28399689 on Build GRCh38
1157773 c.1326G>C p.L442L Chr.17: 28399707-28399707 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781810948 Chr.17: 28399615-28399615 on Build GRCh38
rs782746393 Chr.17: 28399612-28399612 on Build GRCh38
rs782584154 Chr.17: 28399756-28399756 on Build GRCh38
rs886052753 Chr.17: 28399652-28399652 on Build GRCh38
rs141934516 Chr.17: 28399733-28399733 on Build GRCh38
rs782337625 Chr.17: 28399635-28399635 on Build GRCh38
rs781844965 Chr.17: 28399726-28399726 on Build GRCh38
rs782433354 Chr.17: 28399737-28399737 on Build GRCh38
rs782350227 Chr.17: 28399573-28399573 on Build GRCh38
rs541930259 Chr.17: 28399648-28399648 on Build GRCh38
rs915523147 Chr.17: 28399624-28399624 on Build GRCh38
rs782088415 Chr.17: 28399691-28399691 on Build GRCh38
rs782585207 Chr.17: 28399557-28399557 on Build GRCh38
rs116665535 Chr.17: 28399577-28399577 on Build GRCh38
rs781855453 Chr.17: 28399625-28399625 on Build GRCh38
rs370669989 Chr.17: 28399688-28399688 on Build GRCh38
rs782535733 Chr.17: 28399627-28399627 on Build GRCh38
rs782060590 Chr.17: 28399579-28399579 on Build GRCh38
rs781994095 Chr.17: 28399562-28399562 on Build GRCh38
rs782273845 Chr.17: 28399757-28399757 on Build GRCh38
rs782258219 Chr.17: 28399634-28399634 on Build GRCh38
rs372717860 Chr.17: 28399760-28399760 on Build GRCh38
rs782807118 Chr.17: 28399591-28399591 on Build GRCh38
rs186143284 Chr.17: 28399670-28399670 on Build GRCh38
rs41297121 Chr.17: 28399596-28399596 on Build GRCh38
rs781904464 Chr.17: 28399592-28399592 on Build GRCh38
rs782633511 Chr.17: 28399630-28399630 on Build GRCh38
rs782316541 Chr.17: 28399552-28399552 on Build GRCh38
rs781826739 Chr.17: 28399605-28399605 on Build GRCh38
rs781799325 Chr.17: 28399716-28399716 on Build GRCh38
rs782615925 Chr.17: 28399645-28399645 on Build GRCh38
rs782079474 Chr.17: 28399566-28399566 on Build GRCh38
rs782007129 Chr.17: 28399689-28399689 on Build GRCh38
rs782368223 Chr.17: 28399561-28399561 on Build GRCh38
rs1042990430 Chr.17: 28399662-28399662 on Build GRCh38
rs782161667 Chr.17: 28399686-28399686 on Build GRCh38
rs782118481 Chr.17: 28399594-28399594 on Build GRCh38
rs886052754 Chr.17: 28399669-28399669 on Build GRCh38
rs782500985 Chr.17: 28399610-28399610 on Build GRCh38
rs369588539 Chr.17: 28399621-28399621 on Build GRCh38
rs200386243 Chr.17: 28399724-28399724 on Build GRCh38
rs782324529 Chr.17: 28399663-28399663 on Build GRCh38
rs782750258 Chr.17: 28399711-28399711 on Build GRCh38

View Full Product Details