Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75003590-75003819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477773 c.331A>T p.K111* Chr.14: 75003597-75003597 on Build GRCh38
5824579 c.367A>T p.S123C Chr.14: 75003633-75003633 on Build GRCh38
7313744 c.420G>T p.L140L Chr.14: 75003686-75003686 on Build GRCh38
5848544 c.344C>T p.S115F Chr.14: 75003610-75003610 on Build GRCh38
7121593 c.353T>G p.L118R Chr.14: 75003619-75003619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745423619 Chr.14: 75003633-75003633 on Build GRCh38
rs760147450 Chr.14: 75003724-75003724 on Build GRCh38
rs372174903 Chr.14: 75003650-75003650 on Build GRCh38
rs367701960 Chr.14: 75003727-75003727 on Build GRCh38
rs772320060 Chr.14: 75003674-75003674 on Build GRCh38
rs1045660845 Chr.14: 75003708-75003708 on Build GRCh38
rs150617429 Chr.14: 75003646-75003646 on Build GRCh38
rs1039929308 Chr.14: 75003643-75003643 on Build GRCh38
rs1024990084 Chr.14: 75003794-75003794 on Build GRCh38
rs374195020 Chr.14: 75003620-75003620 on Build GRCh38
rs762017222 Chr.14: 75003673-75003673 on Build GRCh38
rs766487534 Chr.14: 75003714-75003714 on Build GRCh38
rs760706892 Chr.14: 75003706-75003706 on Build GRCh38
rs201622896 Chr.14: 75003667-75003667 on Build GRCh38
rs777077732 Chr.14: 75003666-75003668 on Build GRCh38
rs376824565 Chr.14: 75003707-75003707 on Build GRCh38
rs769455036 Chr.14: 75003638-75003638 on Build GRCh38
rs368343040 Chr.14: 75003621-75003621 on Build GRCh38
rs780945717 Chr.14: 75003745-75003745 on Build GRCh38
rs175041 Chr.14: 75003743-75003743 on Build GRCh38
rs779349624 Chr.14: 75003611-75003611 on Build GRCh38
rs929643497 Chr.14: 75003669-75003669 on Build GRCh38
rs751159755 Chr.14: 75003612-75003612 on Build GRCh38
rs773252033 Chr.14: 75003689-75003689 on Build GRCh38
rs200642949 Chr.14: 75003648-75003648 on Build GRCh38
rs748297529 Chr.14: 75003675-75003676 on Build GRCh38
rs201912801 Chr.14: 75003671-75003671 on Build GRCh38
rs754240366 Chr.14: 75003715-75003715 on Build GRCh38
rs753157137 Chr.14: 75003739-75003739 on Build GRCh38

View Full Product Details