Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7702187-7702456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_011523952.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985003 c.1001G>T p.S334I Chr.17: 7702389-7702389 on Build GRCh38
4960820 c.1037G>A p.G346D Chr.17: 7702425-7702425 on Build GRCh38
6823335 c.1002C>T p.S334S Chr.17: 7702390-7702390 on Build GRCh38
473439 c.1024T>C p.C342R Chr.17: 7702412-7702412 on Build GRCh38
6267375 c.1057G>T p.A353S Chr.17: 7702445-7702445 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1052383910 Chr.17: 7702249-7702249 on Build GRCh38
rs574637496 Chr.17: 7702227-7702228 on Build GRCh38
rs377611540 Chr.17: 7702427-7702427 on Build GRCh38
rs995586153 Chr.17: 7702340-7702340 on Build GRCh38
rs770582241 Chr.17: 7702298-7702298 on Build GRCh38
rs138784430 Chr.17: 7702423-7702423 on Build GRCh38
rs73248509 Chr.17: 7702234-7702234 on Build GRCh38
rs778192810 Chr.17: 7702422-7702422 on Build GRCh38
rs903442504 Chr.17: 7702228-7702228 on Build GRCh38
rs908589383 Chr.17: 7702312-7702312 on Build GRCh38
rs971585446 Chr.17: 7702276-7702276 on Build GRCh38
rs764119462 Chr.17: 7702327-7702327 on Build GRCh38
rs773825543 Chr.17: 7702307-7702307 on Build GRCh38
rs889310448 Chr.17: 7702252-7702252 on Build GRCh38
rs1006575779 Chr.17: 7702265-7702265 on Build GRCh38
rs748509622 Chr.17: 7702405-7702405 on Build GRCh38
rs759405865 Chr.17: 7702313-7702313 on Build GRCh38
rs767255046 Chr.17: 7702314-7702314 on Build GRCh38
rs770311262 Chr.17: 7702311-7702311 on Build GRCh38
rs991571322 Chr.17: 7702429-7702429 on Build GRCh38
rs983356988 Chr.17: 7702286-7702286 on Build GRCh38
rs756619906 Chr.17: 7702408-7702408 on Build GRCh38
rs146558070 Chr.17: 7702363-7702363 on Build GRCh38
rs192994436 Chr.17: 7702336-7702336 on Build GRCh38
rs759174078 Chr.17: 7702435-7702435 on Build GRCh38
rs758704444 Chr.17: 7702364-7702364 on Build GRCh38
rs111697163 Chr.17: 7702343-7702343 on Build GRCh38
rs376918892 Chr.17: 7702321-7702321 on Build GRCh38
rs981268255 Chr.17: 7702369-7702369 on Build GRCh38
rs781468333 Chr.17: 7702396-7702396 on Build GRCh38
rs766656480 Chr.17: 7702266-7702266 on Build GRCh38
rs371006232 Chr.17: 7702428-7702428 on Build GRCh38
rs538474084 Chr.17: 7702424-7702424 on Build GRCh38
rs773916846 Chr.17: 7702338-7702338 on Build GRCh38
rs369984114 Chr.17: 7702371-7702371 on Build GRCh38
rs780213284 Chr.17: 7702404-7702404 on Build GRCh38

View Full Product Details