Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42724446-42724673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
UniGene
Hs.599232
Gene Chromosome Location
Chr.15: 42723557-42737219 on Build GRCh38
Transcripts
NM_138477.2 XM_005254176.4 XM_011521270.2 XM_011521271.2 XM_011521274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601214 c.3613C>T p.L1205L Chr.15: 42724562-42724562 on Build GRCh38
961837 c.3575T>G p.L1192* Chr.15: 42724600-42724600 on Build GRCh38
1608383 c.3593T>G p.L1198R Chr.15: 42724582-42724582 on Build GRCh38
6032824 c.3586T>A p.S1196T Chr.15: 42724589-42724589 on Build GRCh38
961836 c.3598C>A p.L1200I Chr.15: 42724577-42724577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939565372 Chr.15: 42724529-42724529 on Build GRCh38
rs192855524 Chr.15: 42724487-42724487 on Build GRCh38
rs768668105 Chr.15: 42724636-42724636 on Build GRCh38
rs368759902 Chr.15: 42724571-42724571 on Build GRCh38
rs573902123 Chr.15: 42724572-42724572 on Build GRCh38
rs375592947 Chr.15: 42724475-42724475 on Build GRCh38
rs745737650 Chr.15: 42724584-42724584 on Build GRCh38
rs766708309 Chr.15: 42724535-42724535 on Build GRCh38
rs375128684 Chr.15: 42724627-42724627 on Build GRCh38
rs543020087 Chr.15: 42724626-42724626 on Build GRCh38
rs11857216 Chr.15: 42724646-42724646 on Build GRCh38
rs374471987 Chr.15: 42724569-42724569 on Build GRCh38
rs138934554 Chr.15: 42724527-42724527 on Build GRCh38
rs77191722 Chr.15: 42724509-42724509 on Build GRCh38
rs866710253 Chr.15: 42724538-42724538 on Build GRCh38
rs771780683 Chr.15: 42724585-42724585 on Build GRCh38
rs758676685 Chr.15: 42724533-42724533 on Build GRCh38
rs935714182 Chr.15: 42724551-42724551 on Build GRCh38
rs537513538 Chr.15: 42724587-42724587 on Build GRCh38
rs867688636 Chr.15: 42724516-42724516 on Build GRCh38
rs370906448 Chr.15: 42724578-42724578 on Build GRCh38
rs979972750 Chr.15: 42724524-42724524 on Build GRCh38
rs772116614 Chr.15: 42724468-42724468 on Build GRCh38
rs746975556 Chr.15: 42724630-42724630 on Build GRCh38
rs915288145 Chr.15: 42724473-42724473 on Build GRCh38
rs753360990 Chr.15: 42724492-42724492 on Build GRCh38
rs764763838 Chr.15: 42724517-42724517 on Build GRCh38
rs992728743 Chr.15: 42724485-42724485 on Build GRCh38
rs917201859 Chr.15: 42724502-42724502 on Build GRCh38
rs558357051 Chr.15: 42724489-42724489 on Build GRCh38
rs201592509 Chr.15: 42724552-42724552 on Build GRCh38
rs371064172 Chr.15: 42724530-42724530 on Build GRCh38
rs755434169 Chr.15: 42724553-42724553 on Build GRCh38
rs760632372 Chr.15: 42724479-42724479 on Build GRCh38

View Full Product Details