Product Details

Assay Reference Genome
Location

Chr.9:132895041 on build GRCh38
Cytoband
9q34.13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
7248
Gene Symbol
TSC1
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
Location
Chr.9:132891348-132945269 on build GRCh38
Assay Gene Location
Within Exon 25
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
TSC1 NM_000368.4 23 4923 NP_000359.1
NM_001162426.1 23 4920 NP_001155898.1
NM_001162427.1 22 4770 NP_001155899.1
XM_005272211.1 23 4968 XP_005272268.1
XM_006717271.1 23 4903 XP_006717334.1
XM_011518979.2 23 4917 XP_011517281.1
XM_017015096.1 24 5382 XP_016870585.1
XM_017015097.1 24 5353 XP_016870586.1
XM_017015098.1 23 4965 XP_016870587.1
XM_017015099.1 22 5061 XP_016870588.1
XM_017015100.1 22 5032 XP_016870589.1
XM_017015101.1 22 5059 XP_016870590.1
AF013168.1 23 4910 AAC51674.1
D87683.1 10 3294 BAA13436.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv831743 Chr9:132820292 - 132982548 on Build GRCh38 Loss MIR548AW GFI1B TSC1 AK8 SPACA9
esv2739138 Chr9:132727483 - 135521553 on Build GRCh38 Deletion MIR3689D2 MIR3689D1 CEL MIR3689B LOC101448202 MRPS2 PPP1R26 RPL7A SNORD36B DBH RNU6ATAC SARDH MIR548AW OBP2B FCN2 WDR5 TSC1 COL5A1-AS1 DBH-AS1 VAV2 C9orf62 STKLD1 SURF2 OLFM1 ADAMTS13 MIR3689A ADAMTSL2 GBGT1 LINC00094 SURF4 TMEM8C CACFD1 LOC101928193 MIR3689C SURF1 REXO4 MIR4669 PPP1R26-AS1 SNORD36C BRD3 RXRA SPACA9 CELP C9orf116 MED22 FCN1 MIR6877 LOC101928525 MIR3689E COL5A1 LOC100996574 SURF6 GTF3C5 LOC100130548 RALGDS LOC401557 MIR3689F FAM163B SNORD24 SLC2A6 LOC105376306 ABO GFI1B SNORD141A SNORD36A AK8 LCN1

View Full Product Details